Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001225.3 | NP_001216.1 | 3 | 379 | |
NM_033306.2 | NP_150649.1 | 3 | 412 | ||
XM_011543019.1 | XP_011541321.1 | 3 | 435 | ||
XM_017018397.1 | XP_016873886.1 | 3 | 379 | ||
GenBank mRNA | AK057094.1 | - | 3 | 1217 | |
AK225751.1 | - | 3 | 350 | ||
AK296081.1 | - | 3 | 356 | ||
AK304222.1 | - | 3 | 362 | ||
BC017839.1 | - | 3 | 345 | 4699374 | |
CN261701.1 | - | 3 | 408 | ||
U25804.1 | - | 3 | 354 | ||
U28014.1 | - | 3 | 343 | ||
U28976.1 | - | 3 | 339 | ||
U28977.1 | - | 3 | 414 | ||
U28978.1 | - | 3 | 412 | ||
U28979.1 | - | 3 | 387 | ||
Z48810.1 | - | 3 | 347 |