Gene Transcripts

Gene Symbol
HS6ST3
Entrez Gene ID
266722
Gene Name
heparan sulfate 6-O-sulfotransferase 3
Gene Aliases
HS6ST-3
Chromosome Location
Chr. 13: 96090634 - 96839562 on Build GRCh38
UniGene ID
Hs.171001
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153456.3 NP_703157.2 2 1489
GenBank mRNA AF539426.1 - 2 1489
AK126947.1 - 2 991
CR749677.1 - 2 1155

View Full Product Details