Gene Transcripts

Gene Symbol
C17orf78
Entrez Gene ID
284099
Gene Name
chromosome 17 open reading frame 78
Gene Aliases
Chromosome Location
Chr. 17: 37375710 - 37392708 on Build GRCh38
UniGene ID
Hs.439154
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321399.1 NP_001308328.1 5 1323
NM_173625.4 NP_775896.3 7 1565
XM_011524647.2 XP_011522949.1 6 1754
XM_011524648.2 XP_011522950.1 6 1722
GenBank mRNA AK096966.1 - 5 1324
BC034672.1 - 7 1508 5184947

View Full Product Details