Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_174978.2 | NP_777638.2 | 4 | 298 | |
XM_011536690.2 | XP_011534992.1 | 5 | 428 | ||
XM_011536703.2 | XP_011535005.1 | 5 | 428 | ||
XM_011536704.1 | XP_011535006.1 | 5 | 429 | ||
XM_017021247.1 | XP_016876736.1 | 5 | 429 | ||
XM_017021248.1 | XP_016876737.1 | 3 | 261 | ||
GenBank mRNA | AK098187.1 | - | 4 | 283 | |
AL832219.1 | - | 4 | 348 | ||
BC125070.1 | - | 4 | 269 | 40117603 |