Gene Transcripts

Gene Symbol
TAAR9
Entrez Gene ID
134860
Gene Name
trace amine associated receptor 9 (gene/pseudogene)
Gene Aliases
TA3, TAR3, TAR9, TRAR3
Chromosome Location
Chr. 6: 132538288 - 132539336 on Build GRCh38
UniGene ID
Hs.434116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175057.3 NP_778227.3 1 98
GenBank mRNA AY183469.1 - 1 96
BC069083.1 - 1 98

View Full Product Details