Gene Transcripts

Gene Symbol
TRIM3
Entrez Gene ID
10612
Gene Name
tripartite motif containing 3
Gene Aliases
BERP, HAC1, RNF22, RNF97
Chromosome Location
Chr. 11: 6448613 - 6474459 on Build GRCh38
UniGene ID
Hs.591992
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001248006.1 NP_001234935.1 2 502
NM_006458.3 NP_006449.2 3 503
NM_033278.3 NP_150594.2 2 338
XM_011519844.1 XP_011518146.1 3 602
XM_011519845.2 XP_011518147.1 2 288
XM_017017098.1 XP_016872587.1 2 314
XM_017017099.1 XP_016872588.1 2 211
GenBank mRNA AF045239.1 - 1 107
AF220020.1 - 1 107
AF220021.1 - 1 107
AF220022.1 - 1 107
AK126685.1 - 2 217
AK291396.1 - 2 502
AK299586.1 - 2 295
AK311127.1 - 2 298
AL360192.1 - 1 82
BC096827.1 - 2 338 6108991
BI916995.1 - 3 442
BU500178.1 - 348
DA089323.1 - 2 213

View Full Product Details