Gene Transcripts
- Gene Symbol
- Nhsl2
- Entrez Gene ID
- 100042480
- Gene Name
- NHS-like 2
- Gene Aliases
- 1110062M06Rik, 6430511F03, AI225852, AW488865, EG621083, ENSMUSG00000073037, Gm10456
- Chromosome Location
- Chr. X: 101848669 - 102092055 on Build GRCm38
- UniGene ID
- Mm.232
- Species
- Mus musculus
View Full Product Details