Gene Transcripts

Gene Symbol
Aim1l
Entrez Gene ID
230806
Gene Name
absent in melanoma 1-like
Gene Aliases
Chromosome Location
Chr. 4: 134060728 - 134092504 on Build GRCm38
UniGene ID
Mm.240080
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162970.1 NP_001156442.1 2 399
GenBank mRNA AK145690.1 - 2 402
BC150738.1 - 1 328 9087649

View Full Product Details