Gene Transcripts

Gene Symbol
Bex6
Entrez Gene ID
328660
Gene Name
brain expressed gene 6
Gene Aliases
B020003O03Rik
Chromosome Location
Chr. 16: 32179800 - 32186944 on Build GRCm38
UniGene ID
Mm.441006
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033539.2 NP_001028711.1 1 38
GenBank mRNA AK157452.1 - 1 39
AY833559.1 - 1 38

View Full Product Details