Gene Transcripts

Gene Symbol
Ankrd63
Entrez Gene ID
383787
Gene Name
ankyrin repeat domain 63
Gene Aliases
Gm1337
Chromosome Location
Chr. 2: 118699103 - 118703963 on Build GRCm38
UniGene ID
Mm.35758
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081971.1 NP_001075440.1 1 2259
GenBank mRNA AK087787.1 - 1 946
AK135101.1 - 1 2159

View Full Product Details