Gene Transcripts

Gene Symbol
Spag11b
Entrez Gene ID
546038
Gene Name
sperm associated antigen 11B
Gene Aliases
EG546038, Ep2c/h, Spag11c/h
Chromosome Location
Chr. 8: 19140759 - 19143010 on Build GRCm38
UniGene ID
Mm.99206
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034905.2 NP_001030077.1 2 923
GenBank mRNA AK136627.1 - 2 924

View Full Product Details