Gene Transcripts

Gene Symbol
Tatdn3
Entrez Gene ID
68972
Gene Name
TatD DNase domain containing 3
Gene Aliases
1500010M24Rik
Chromosome Location
Chr. 1: 191045826 - 191063010 on Build GRCm38
UniGene ID
Mm.237188
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163422.1 NP_001156894.1 7 544
GenBank mRNA AK005193.1 - 7 515

View Full Product Details