Gene Transcripts

Gene Symbol
Arpc1b
Entrez Gene ID
54227
Gene Name
actin related protein 2/3 complex, subunit 1B
Gene Aliases
Chromosome Location
Chr. 12: 11252300 - 11265849 on Build Rnor_6.0
UniGene ID
Rn.2090
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019289.2 NP_062162.1 3 188
GenBank mRNA AF083269.1 - 2 101
BC062027.1 - 3 188 5622004
FQ228791.1 - 3 159
FQ228917.1 - 3 204

View Full Product Details