Gene Transcripts

Gene Symbol
Arl16
Entrez Gene ID
70317
Gene Name
ADP-ribosylation factor-like 16
Gene Aliases
2600005N12Rik
Chromosome Location
Chr. 11: 120464326 - 120467600 on Build GRCm38
UniGene ID
Mm.485046
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_197995.2 NP_932112.2 3 380
GenBank mRNA AK011161.1 - 2 176
AK038349.1 - 3 252
AK140077.1 - 2 384
BC009089.1 - 3 221
BC139260.1 - 3 251 8862282
BC139262.1 - 3 251 8862284
BF472168.1 - 3 387

View Full Product Details