Gene Transcripts

Gene Symbol
Aqp6
Entrez Gene ID
29170
Gene Name
aquaporin 6, kidney specific
Gene Aliases
Chromosome Location
Chr. 7: 141258585 - 141261518 on Build Rnor_6.0
UniGene ID
Rn.48667
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022181.1 NP_071517.1 4 902
GenBank mRNA AF083879.1 - 4 902
L28113.1 - 4 930

View Full Product Details