Gene Transcripts
- Gene Symbol
- Pdx1
- Entrez Gene ID
- 29535
- Gene Name
- pancreatic and duodenal homeobox 1
- Gene Aliases
- Idx1, Ipf1, Stf1
- Chromosome Location
- Chr. 12: 9496044 - 9501211 on Build Rnor_6.0
- UniGene ID
- Rn.54603
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022852.3 |
NP_074043.3
|
2 |
540 |
|
GenBank mRNA |
U04833.1 |
- |
2 |
540 |
|
View Full Product Details