Gene Transcripts

Gene Symbol
Cgrrf1
Entrez Gene ID
116679
Gene Name
cell growth regulator with ring finger domain 1
Gene Aliases
Cgr19
Chromosome Location
Chr. 15: 23619176 - 23639622 on Build Rnor_6.0
UniGene ID
Rn.87514
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053899.1 NP_446351.1 4 654
GenBank mRNA BC078763.1 - 3 800
FQ214200.1 - 3 695
FQ218613.1 - 4 669
U66471.1 - 4 654

View Full Product Details