Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199770.1 | NP_001186699.1 | 5 | 1355 | |
NM_014562.3 | NP_055377.1 | 5 | 1355 | ||
NR_130153.1 | 5 | 1470 | |||
GenBank mRNA | AK095680.1 | - | 5 | 1327 | |
AK297308.1 | - | 5 | 1441 | ||
BC007621.1 | - | 5 | 1299 | 3355563 |