Gene Transcripts

Gene Symbol
Carhsp1
Entrez Gene ID
260416
Gene Name
calcium regulated heat stable protein 1
Gene Aliases
Crhsp-24, Crhsp24
Chromosome Location
Chr. 10: 7041033 - 7055107 on Build Rnor_6.0
UniGene ID
Rn.14584
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152790.2 NP_690003.1 4 371
XM_006245750.2 XP_006245812.1 4 1154
XM_006245753.2 XP_006245815.1 4 525
XM_008767479.1 XP_008765701.1 4 848
GenBank mRNA AF115346.1 - 3 341
BC071173.1 - 4 371 7099252

View Full Product Details