Gene Transcripts

Gene Symbol
Sec11c
Entrez Gene ID
266758
Gene Name
SEC11 homolog C (S. cerevisiae)
Gene Aliases
Sec11l3, Spc21
Chromosome Location
Chr. 18: 61490051 - 61506393 on Build Rnor_6.0
UniGene ID
Rn.16883
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153628.2 NP_705892.1 3 364
GenBank mRNA AB022714.2 - 3 365

View Full Product Details