Gene Transcripts
- Gene Symbol
- Cst12
- Entrez Gene ID
- 266776
- Gene Name
- cystatin 12
- Gene Aliases
- Chromosome Location
- Chr. 3: 143119697 - 143123557 on Build Rnor_6.0
- UniGene ID
- Rn.58124
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153734.1 |
NP_714956.1
|
2, 3 |
362 |
|
GenBank mRNA |
AF440736.1 |
- |
2, 3 |
362 |
|
View Full Product Details