Gene Transcripts

Gene Symbol
Bpifa1
Entrez Gene ID
246238
Gene Name
BPI fold containing family A, member 1
Gene Aliases
Plunc
Chromosome Location
Chr. 3: 149624867 - 149630610 on Build Rnor_6.0
UniGene ID
Rn.19272
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172031.1 NP_742028.1 5 621
GenBank mRNA AF393750.1 - 5 621

View Full Product Details