Gene Transcripts
- Gene Symbol
- Htr4
- Entrez Gene ID
- 15562
- Gene Name
- 5 hydroxytryptamine (serotonin) receptor 4
- Gene Aliases
- 5-HT-4, 5-HT4, 5-HT<4L>, 5HTR4
- Chromosome Location
- Chr. 18: 62322160 - 62498026 on Build GRCm38
- UniGene ID
- Mm.20440
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008313.4 |
NP_032339.2
|
3, 4 |
363 |
|
GenBank mRNA |
AJ011369.1 |
- |
2, 3 |
153 |
|
AK141587.1 |
- |
3, 4 |
307 |
|
AK141625.1 |
- |
3, 4 |
364 |
|
BC132189.1 |
- |
2, 3 |
192 |
|
BC132191.1 |
- |
2, 3 |
192 |
|
BC145159.1 |
- |
2, 3 |
192 |
|
BC145161.1 |
- |
2, 3 |
192 |
|
JN418927.1 |
- |
3, 4 |
294 |
|
JN418928.1 |
- |
3, 4 |
280 |
|
JN418930.1 |
- |
2, 3 |
169 |
|
Y09585.1 |
- |
2, 3 |
153 |
|
Y09587.1 |
- |
2, 3 |
153 |
|
Y09588.1 |
- |
2, 3 |
153 |
|
View Full Product Details