Gene Transcripts
- Gene Symbol
- Spt1
- Entrez Gene ID
- 20770
- Gene Name
- salivary protein 1
- Gene Aliases
- Spt-1
- Chromosome Location
- Chr. 15: 103895855 - 103899312 on Build GRCm38
- UniGene ID
- Mm.1280
- Species
- Mus musculus
- Gene Symbol
- Mucl1
- Entrez Gene ID
- 20771
- Gene Name
- mucin-like 1
- Gene Aliases
- Spt-2, Spt2
- Chromosome Location
- Chr. 15: 103751923 - 103755498 on Build GRCm38
- UniGene ID
- Mm.30276
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC091766.1 |
- |
|
173 |
|
View Full Product Details