Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010798.2 | NP_034928.1 | 2 | 228 | |
GenBank mRNA | AK008939.1 | - | 2 | 222 | |
AK010691.1 | - | 2 | 199 | ||
AK013328.1 | - | 2 | 222 | ||
AK152211.1 | - | 2 | 209 | ||
BC024895.1 | - | 2 | 208 | 3978685 | |
BC086928.1 | - | 2 | 198 | 6762214 | |
BC100605.1 | - | 2 | 198 | 30118109 | |
L02913.1 | - | 2 | 201 | ||
L07607.1 | - | 2 | 131 | ||
L10613.1 | - | 2 | 227 | ||
Z23048.1 | - | 2 | 148 |