Gene Transcripts
- Gene Symbol
- Map6
- Entrez Gene ID
- 17760
- Gene Name
- microtubule-associated protein 6
- Gene Aliases
- 2810411E12Rik, Map-6, Mtap6, STOP
- Chromosome Location
- Chr. 7: 99267447 - 99337137 on Build GRCm38
- UniGene ID
- Mm.154087
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001043355.2 |
NP_001036820.2
|
1, 2 |
976 |
|
NM_001048167.1 |
NP_001041632.1
|
1, 2 |
1875 |
|
NM_010837.3 |
NP_034967.2
|
1, 2 |
1875 |
|
GenBank mRNA |
AK018193.2 |
- |
1, 2 |
1355 |
|
AK087413.1 |
- |
1, 2 |
1838 |
|
AK154455.1 |
- |
1, 2 |
948 |
|
AK214985.1 |
- |
1, 2 |
278 |
|
BC053039.1 |
- |
1, 2 |
1876 |
5709455 |
BY259340.1 |
- |
1, 2 |
198 |
|
Y16032.1 |
- |
1, 2 |
916 |
|
View Full Product Details