Gene Transcripts

Gene Symbol
Pon1
Entrez Gene ID
18979
Gene Name
paraoxonase 1
Gene Aliases
Pon
Chromosome Location
Chr. 6: 5168090 - 5193987 on Build GRCm38
UniGene ID
Mm.237657
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011134.3 NP_035264.2 9 1438
GenBank mRNA BC012706.1 - 9 1311
U32684.1 - 9 1320
U72636.1 - 9 1320

View Full Product Details