Gene Transcripts

Gene Symbol
Sphk1
Entrez Gene ID
20698
Gene Name
sphingosine kinase 1
Gene Aliases
1110006G24Rik, Sk1, Spk1
Chromosome Location
Chr. 11: 116529817 - 116536675 on Build GRCm38
UniGene ID
Mm.20944
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172472.1 NP_001165943.1 3 579
NM_001172473.1 NP_001165944.1 3 465
NM_001172475.1 NP_001165946.1 3 504
NM_011451.3 NP_035581.1 1 236
NM_025367.6 NP_079643.2 3 462
GenBank mRNA AF068748.1 - 3 504
AF068749.1 - 1 236
AF415213.1 - 1 134
AK028667.1 - 1 2526
AK136289.1 - 2 250
AK155332.1 - 3 564
AK160900.1 - 3 455
AK197304.1 - 2 149
BC011432.1 - 1 437
BC037710.1 - 3 377 4221357

View Full Product Details