Gene Transcripts

Gene Symbol
Actn3
Entrez Gene ID
11474
Gene Name
actinin alpha 3
Gene Aliases
Chromosome Location
Chr. 19: 4861216 - 4877909 on Build GRCm38
UniGene ID
Mm.5316
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013456.2 NP_038484.1 3 349
GenBank mRNA AF093775.1 - 3 324
BC111890.1 - 3 315 40060598
BY132390.1 - 3 349

View Full Product Details