Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013877.3 | NP_038905.1 | 3 | 287 | |
GenBank mRNA | AB079605.1 | - | 2 | 103 | |
AF169161.1 | - | 3 | 248 | ||
AK044544.1 | - | 3 | 288 | ||
AK087416.1 | - | 3 | 148 | ||
BC018253.1 | - | 3 | 310 | 4504283 |