Gene Transcripts

Gene Symbol
Ddah2
Entrez Gene ID
51793
Gene Name
dimethylarginine dimethylaminohydrolase 2
Gene Aliases
1110003M04Rik, AU019324, AW413173, DDAH-2, DDAHII, Ddah, G6a, NG30
Chromosome Location
Chr. 17: 35059035 - 35062099 on Build GRCm38
UniGene ID
Mm.1457
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190449.1 NP_001177378.1 7 1272
NM_016765.3 NP_058045.1 7 1246
GenBank mRNA AF004106.1 - 7 1180
BC003328.1 - 7 1222 3158175

View Full Product Details