Gene Transcripts

Gene Symbol
KCNA10
Entrez Gene ID
3744
Gene Name
potassium voltage-gated channel subfamily A member 10
Gene Aliases
Kcn1, Kv1.8
Chromosome Location
Chr. 1: 110517217 - 110519175 on Build GRCh38
UniGene ID
Hs.248140
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005549.2 NP_005540.1 1 939
GenBank mRNA BC074990.2 - 1 581

View Full Product Details