Gene Transcripts
- Gene Symbol
- Arhgap31
- Entrez Gene ID
- 12549
- Gene Name
- Rho GTPase activating protein 31
- Gene Aliases
- 5830477L08Rik, AU041750, Cdgap, D330026I07Rik
- Chromosome Location
- Chr. 16: 38598343 - 38713035 on Build GRCm38
- UniGene ID
- Mm.370848
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020260.2 |
NP_064656.2
|
2, 3 |
573 |
|
GenBank mRNA |
AF151363.1 |
- |
2, 3 |
214 |
|
AK018057.1 |
- |
2, 3 |
439 |
|
AK138417.1 |
- |
2, 3 |
813 |
|
AK149975.1 |
- |
2, 3 |
409 |
|
AK150109.1 |
- |
2, 3 |
409 |
|
AK155729.1 |
- |
2, 3 |
393 |
|
AK170685.1 |
- |
2, 3 |
393 |
|
BB865916.1 |
- |
2, 3 |
575 |
|
BC138870.1 |
- |
2, 3 |
378 |
8861892 |
View Full Product Details