Gene Transcripts

Gene Symbol
Anapc5
Entrez Gene ID
59008
Gene Name
anaphase-promoting complex subunit 5
Gene Aliases
2510006G12Rik, AA408751, AA536819, AA986414, APC5, Anpc5
Chromosome Location
Chr. 5: 122787459 - 122821342 on Build GRCm38
UniGene ID
Mm.332667
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042491.2 NP_001035956.1 6 869
NM_001289517.1 NP_001276446.1 6 869
NM_001289518.1 NP_001276447.1 6 869
NM_001289519.1 NP_001276448.1 6 869
NM_001289520.1 NP_001276449.1 6 869
NM_021505.3 NP_067480.2 6 869
GenBank mRNA AB041593.1 - 6 812
AK003821.1 - 3 262
AK031432.1 - 6 794
AK049719.1 - 6 823
AK075969.1 - 6 808
AK088327.1 - 6 832
AK147951.1 - 6 808
AK148679.1 - 6 824
AK151314.1 - 6 815
AK151905.1 - 6 815
AK152528.1 - 6 785
AK152981.1 - 6 815
AK153373.1 - 6 815
AK153422.1 - 6 815
AK159204.1 - 6 815
AK159713.1 - 6 808
AK159916.1 - 6 808
AK167561.1 - 6 808
AK168135.1 - 6 793
AK168737.1 - 6 809
AK168814.1 - 6 808
AK169196.1 - 6 832
AK215226.1 - 1 63
BC010339.1 - 6 785 2648173
BC046566.1 - 6 848 2650790
BC046804.1 - 6 806 6404923

View Full Product Details