Gene Transcripts

Gene Symbol
AQP9
Entrez Gene ID
366
Gene Name
aquaporin 9
Gene Aliases
AQP-9, HsT17287, SSC1, T17287
Chromosome Location
Chr. 15: 58138169 - 58185911 on Build GRCh38
UniGene ID
Hs.104624
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB008775.1 - 6 1058
AF016495.1 - 6 1070
AK300055.1 - 6 817
AK313214.1 - 6 1137
BC026258.1 - 6 1027 4734559
CR542027.1 - 6 842

View Full Product Details