Gene Transcripts

Gene Symbol
Mpnd
Entrez Gene ID
68047
Gene Name
MPN domain containing
Gene Aliases
E130307M08Rik
Chromosome Location
Chr. 17: 56009157 - 56016790 on Build GRCm38
UniGene ID
Mm.358710
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026530.5 NP_080806.4 3 480
GenBank mRNA AK012128.1 - 2 435
AK146973.1 - 3 465
AK160356.1 - 3 449
AK172381.1 - 2 223
AK199908.1 - 2 198
BC017625.1 - 2 429

View Full Product Details