Gene Transcripts

Gene Symbol
PEX12
Entrez Gene ID
5193
Gene Name
peroxisomal biogenesis factor 12
Gene Aliases
PAF-3, PBD3A
Chromosome Location
Chr. 17: 35574795 - 35578637 on Build GRCh38
UniGene ID
Hs.591190
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000286.2 NP_000277.1 3 2140
GenBank mRNA AB004546.1 - 3 1709
BC031085.1 - 3 2044 5296541
U91521.1 - 3 2071

View Full Product Details