Gene Transcripts

Gene Symbol
Snf8
Entrez Gene ID
27681
Gene Name
SNF8, ESCRT-II complex subunit, homolog (S. cerevisiae)
Gene Aliases
D11Moh34
Chromosome Location
Chr. 11: 96034890 - 96047430 on Build GRCm38
UniGene ID
Mm.372235
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033568.2 NP_291046.1 4, 5 454
GenBank mRNA AK013058.1 - 4, 5 456
AK020203.1 - 2, 3 144
BC003938.1 - 4, 5 426 3486703

View Full Product Details