Gene Transcripts
- Gene Symbol
- Scgb3a2
- Entrez Gene ID
- 117158
- Gene Name
- secretoglobin, family 3A, member 2
- Gene Aliases
- LuLeu1, Pnsp1, UGRP1, Utgrp1
- Chromosome Location
- Chr. 18: 43764281 - 43767399 on Build GRCm38
- UniGene ID
- Mm.210437
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF274961.1 |
- |
2 |
458 |
|
View Full Product Details