Gene Transcripts

Gene Symbol
Vmn1r8
Entrez Gene ID
171205
Gene Name
vomeronasal 1 receptor 8
Gene Aliases
V1rc32
Chromosome Location
Chr. 6: 57032341 - 57037125 on Build GRCm38
UniGene ID
Mm.222638
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134187.3 NP_598948.1 1 956
GenBank mRNA AF394951.1 - 1 956

View Full Product Details