Gene Transcripts

Gene Symbol
Ubfd1
Entrez Gene ID
28018
Gene Name
ubiquitin family domain containing 1
Gene Aliases
AI467302, D7Wsu105e, D7Wsu128e, Ubph
Chromosome Location
Chr. 7: 122067198 - 122082199 on Build GRCm38
UniGene ID
Mm.282122
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138589.2 NP_613055.2 3 648
GenBank mRNA AK011862.1 - 3 404
AK017851.1 - 3 425
AK033428.1 - 2 1427
AK077395.1 - 3 671
AK135258.1 - 3 406
AK140891.1 - 3 407
BC011313.1 - 3 651 3499738

View Full Product Details