Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_139236.3 | NP_631982.2 | 3 | 440 | |
GenBank mRNA | AF361077.1 | - | 3 | 389 | |
AF361078.1 | - | 3 | 389 | ||
AK089138.1 | - | 3 | 360 | ||
AK155818.1 | - | 3 | 379 | ||
AK166559.1 | - | 3 | 366 | ||
AK166650.1 | - | 3 | 366 | ||
AK171163.1 | - | 3 | 377 | ||
AK171421.1 | - | 3 | 367 | ||
AK191861.1 | - | 3 | 296 | ||
AK195706.1 | - | 3 | 296 | ||
AK197974.1 | - | 3 | 296 | ||
AK198310.1 | - | 3 | 296 | ||
AK199838.1 | - | 3 | 297 | ||
AK202668.1 | - | 3 | 296 | ||
AK204352.1 | - | 3 | 299 | ||
AK209500.1 | - | 3 | 296 | ||
BC059820.1 | - | 3 | 358 | 6827596 | |
BC060189.1 | - | 3 | 368 |