Gene Transcripts

Gene Symbol
Taf6l
Entrez Gene ID
225895
Gene Name
TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor
Gene Aliases
2810417N14Rik, BB223262, C530024J06Rik, Paf65a
Chromosome Location
Chr. 19: 8774354 - 8786417 on Build GRCm38
UniGene ID
Mm.220886
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177798.1 NP_001171269.1 10 1233
NM_146092.2 NP_666204.2 10 1070
GenBank mRNA AK077149.1 - 10 1234
AK144881.1 - 9 1330
AK209124.1 - 2 141
BC020299.1 - 10 1043
BC028647.1 - 10 1017 1244243

View Full Product Details