Gene Transcripts

Gene Symbol
Zfand3
Entrez Gene ID
21769
Gene Name
zinc finger, AN1-type domain 3
Gene Aliases
AW539211, TEG-27, Tex27
Chromosome Location
Chr. 17: 30005039 - 30210020 on Build GRCm38
UniGene ID
Mm.220972
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148926.2 NP_683728.1 3, 4 450
GenBank mRNA AK082915.1 - 2, 3 446
BC034396.1 - 3, 4 370 3707476
BC083124.1 - 3, 4 450 6334368

View Full Product Details