Gene Transcripts

Gene Symbol
Tor2a
Entrez Gene ID
30933
Gene Name
torsin family 2, member A
Gene Aliases
Prosalusin
Chromosome Location
Chr. 2: 32757231 - 32762244 on Build GRCm38
UniGene ID
Mm.200695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152800.3 NP_690013.1 5 1190
GenBank mRNA AK003976.1 - 1 378
AK162719.1 - 5 1134
AK170979.1 - 4 2369
BC003466.1 - 5 1175 2649388
BC023085.1 - 3 689
BC024469.1 - 5 1153 4976022
BY579003.1 - 1 48

View Full Product Details