Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289722.1 | NP_001276651.1 | 8 | 4347 | |
NM_001289723.1 | NP_001276652.1 | 8 | 4312 | ||
NM_153138.4 | NP_694778.1 | 8 | 4342 | ||
GenBank mRNA | AK156547.1 | - | 8 | 4349 | |
BB740185.1 | - | 1 | 273 | ||
BC049788.1 | - | 8 | 4265 | 6514406 |