Gene Transcripts

Gene Symbol
U2af1l4
Entrez Gene ID
233073
Gene Name
U2 small nuclear RNA auxiliary factor 1-like 4
Gene Aliases
AA407033, AF419339, AI451269, AW553050, U2af26
Chromosome Location
Chr. 7: 30563340 - 30565364 on Build GRCm38
UniGene ID
Mm.34790
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170760.3 NP_739566.1 4 243
GenBank mRNA AF419339.1 - 4 227
AK180639.1 - 1 15
BC051477.1 - 4 251
BC060972.1 - 4 243 6593404

View Full Product Details