Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172577.3 | NP_766165.2 | 1, 2 | 608 | |
GenBank mRNA | AK036988.1 | - | 1, 2 | 348 | |
AK041694.1 | - | 1, 2 | 609 | ||
AK045180.1 | - | 1, 2 | 434 | ||
AK162379.1 | - | 1, 2 | 435 | ||
AK167535.1 | - | 1, 2 | 316 | ||
BC057980.1 | - | 1, 2 | 432 | 5042653 |