Gene Transcripts

Gene Symbol
ADSS
Entrez Gene ID
159
Gene Name
adenylosuccinate synthase
Gene Aliases
ADEH, ADSS 2
Chromosome Location
Chr. 1: 244408492 - 244452134 on Build GRCh38
UniGene ID
Hs.498313
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126.3 NP_001117.2 2 512
XM_011544108.2 XP_011542410.1 2 1184
GenBank mRNA AK025514.1 - 2 259
AK303886.1 - 3 224
AK308749.1 - 3 597
BC012356.1 - 2 297 4635347
BG700800.1 - 2 491

View Full Product Details