Gene Transcripts

Gene Symbol
Klri2
Entrez Gene ID
320407
Gene Name
killer cell lectin-like receptor family I member 2
Gene Aliases
A530090P03Rik
Chromosome Location
Chr. 6: 129729041 - 129740484 on Build GRCm38
UniGene ID
Mm.123265
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177155.4 NP_796129.2 6 2182
GenBank mRNA AK040915.1 - 6 2169

View Full Product Details